JALAS® TIO 2068 ESD Safety Shoes
Supplier: EJENDALS
Certificates
About this item
A soft and water repellent safety shoe with high-quality protection. Ultimate softness and comfort thanks to the soft and stable TIO (Three In One) sole that prevents long term injuries to the feet and back.
- ESD
- Water repellent
- Oil-resistant outsole
- Ergothan Shock Absorption System
- Anti-static properties
- Aluminium toecap
- Nail protection in plasma-treated composite (PTC) textile
The Tri-Panel BOA® Fit system enables a soft and perfect fit for individually shaped feet. The shoe has an aluminium toe cap and soft textile nail-penetration insert. The new three in one sole technology is developed by Jalas.
Prevents risks of toe injuries, nail penetration, antistatic, slipping. Primary environments of use are outdoors, indoors, all-year use, environments with risk for outsole penetration, slippery environments, dry environments, clean environments, warm environments.
Certifications: EN ISO 20345:2011; S3, SRC.