Order Entry
Germany
ContactUsLinkComponent
SamplePro Needle
SamplePro Needle
Catalog # MSPPH160071
SamplePro Needle
Catalog # MSPPH160071

Specifications

  • Ext.Ø:
    0,50 mm
  • Length:
    130 mm
  • Pk:
    1

Specifications

About this item

Non-septum piercing and MATCH tubes even below 3,0 mm outer diameter.

Needle Mounting Adapter

Product Information