ContactUsLinkComponent
Gasket for Flange with Groove
Gasket for Flange with Groove
Catalog # HWSA3395100
Supplier:  HWS LABORTECHNIK
Gasket for Flange with Groove
Catalog # HWSA3395100
Supplier:  HWS LABORTECHNIK
Specifications
  • Description:
    Grrove centering seal, DN100S
  • Pk:
    1
Specifications
About this item
HWS-Gasket in PTFE with locating lip for flange with groove.