Order Entry
Germany
Orders LinkContactUsLinkComponent
Double-stranded RNA (dsRNA) ELISA kit

Double-stranded RNA (dsRNA) ELISA kit

200 AU
Catalog # ABNOKA6886.200
Supplier:   Abnova
Double-stranded RNA (dsRNA) ELISA kit
Catalog # ABNOKA6886.200
Supplier:  Abnova
CAS Number:  
Double-stranded RNA (dsRNA) ELISA kit

200 AU

CAS Number:  
Price
1 370,00 €
Log in for availability
1 * 200 LE
Product Details & Documents

Double-stranded RNA (dsRNA) ELISA Kit is used for sensitive and selective detection of dsRNA molecules.

Specifications

Specifications

  • Pk:
    200 AU
  • Cat. No.:
    ABNOKA6886.200
  • Target protein:
    Double-stranded RNA

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.