Order Entry
Germany
ContactUsLinkComponent
The IgA (Cat) ELISA kit
The IgA (Cat) ELISA kit
Catalog # ABNOKA6753.1KIT
Supplier:  Abnova
CAS Number:  
The IgA (Cat) ELISA kit
Catalog # ABNOKA6753.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Cat
  • Target protein:
    IgA

Specifications

About this item

The IgA (Cat) ELISA Kit is used for immunoperoxidase Assay for Determination of IgA in Cat Samples.