Order Entry
Germany
ContactUsLinkComponent
The IgM (Monkey) ELISA kit
The IgM (Monkey) ELISA kit
Catalog # ABNOKA6751.1KIT
Supplier:  Abnova
CAS Number:  
The IgM (Monkey) ELISA kit
Catalog # ABNOKA6751.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Monkey
  • Target protein:
    IgM

Specifications

About this item

The IgM (Monkey) ELISA Kit is used for determination of IgM in old world monkey serum or plasma.