IgG (Sheep) ELISA kit
Catalog # ABNOKA6765.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Sheep
- Target protein:IgG
- Assay Principle:Quantitative
Specifications
About this item
IgG (Sheep) ELISA Kit is for the quantitative measurement of sheep IgG in serum or plasma.