The DNP (Rat) IgM ELISA kit
Catalog # ABNOKA6742.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Rat
- Target protein:DNP
Specifications
About this item
The DNP (Rat) IgM ELISA Kit is based on a solid phase enzymelinked immunosorbent assay (ELISA). The assay uses DNP-BSA as the capture antigen (coated on the microtiter wells) and HRP conjugated anti-rat IgM antibodies for detection.