Order Entry
Germany
ContactUsLinkComponent
Herpes Simplex Virus I+II IgG ELISA kit
Herpes Simplex Virus I+II IgG ELISA kit
Catalog # ABNOKA6734.1KIT
Supplier:  Abnova
CAS Number:  
Herpes Simplex Virus I+II IgG ELISA kit
Catalog # ABNOKA6734.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    HSV1/2
  • Target protein:
    IgG
  • Assay Principle:
    Qualitative

Specifications

About this item

Herpes Simplex Virus I+II IgG ELISA Kit is a qualitative determination of IgG class antibodies against Herpes Simplex Virus 1+2 (HSV) in human serum or plasma.