Cytomegalovirus IgG ELISA kit
Catalog # ABNOKA6724.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Cytomegalovirus
- Target protein:IgG
- Assay Principle:Qualitative
Specifications
About this item
Cytomegalovirus IgG ELISA Kit is a qualitative determination of IgG class antibodies against Cytomegalovirus (CMV) in human serum or plasma.