LH (Human) ELISA kit
Catalog # ABNOKA6705.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:LH
- Assay Principle:Quantitative
Specifications
About this item
LH (Human) ELISA Kit is an enzyme immunoassay for the quantitative measurement of luteinizing hormone in serum.