Order Entry
Germany
ContactUsLinkComponent
 
AFP (Human) ELISA kit
Catalog # ABNOKA6713.1KIT
Supplier:  Abnova
CAS Number:  
AFP (Human) ELISA kit
Catalog # ABNOKA6713.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    AFP
  • Assay Principle:
    Quantitative

Specifications

About this item

AFP (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of alpha-fetoprotein in serum.