Tetracycline ELISA kit
Catalog # ABNOKA6683.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Target protein:Tetracycline
- Assay Principle:Quantitative
Specifications
About this item
Tetracycline ELISA Kit is a quantitative determination of tetracyclines in muscle, liver, egg, honey, urine.