Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Rabbit LEP (from E. coli)
Recombinant Rabbit LEP (from E. coli)
Recombinant Rabbit LEP (from E. coli)
Catalog # ABNOP8890.100UG
Supplier:
Abnova
CAS Number:
Expand image
Recombinant Rabbit LEP (from E. coli)
Catalog # ABNOP8890.100UG
Supplier:
Abnova
CAS Number:
Specifications
Pk:
100 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Rabbit
Protein/peptide name:
LEP
Specifications
About this item
Product Details
Product Details