Recombinant Human ADIPOQ (from HEK293 cells), FLAG-tag
Catalog # ABNOP8386.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:FLAG-tag
- Gene ID:Q15848
- Protein/peptide name:ADIPOQ