Order Entry
Germany
ContactUsLinkComponent
Recombinant Human TIGIT, His-tag
Recombinant Human TIGIT, His-tag
  ABNOP7601.50UG
 :  Abnova
 :  
Recombinant Human TIGIT, His-tag
  ABNOP7601.50UG
 :  Abnova
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    Q495A1
  • Protein/peptide name:
    TIGIT
  • Sequence:
    22a.a.-141a.a.