Versilon™ PTFE Tubings
Versilon™ PTFE Flexible Tubing from Saint-Gobain has the highest working temperature and chemical resistance of any fluoropolymer tubing available today.
- Fully fluorinated fluoropolymer tubing
- Opaque/translucent colour, other colours available on request
- Highest working temperature (up to 260 °C) out of all fluoropolymer tubing
- Autoclavable
This tubing is ideal for use in general chemical transfer applications such as delivery of natural gas and mineral oils, toxic gas monitoring, and paint, varnish, or adhesive delivery lines. It also serves numerous applications in the automotive, electrical, and appliance markets to insulate wire and cable. It is ideal for applications such as general chemical applications, automotive, electrical, and appliance markets.
Hardness: 50 to 60 (shore D)
- Catalog No:
- VERNA210040010010B
- VERNA210030010010B
- VERNA210048008010A
- VERNA210059010010A
- VERNA200010005010A
- VERNA210025007510B
- Model:
- VERNA210040010010B
- VERNA210030010010B
- VERNA210048008010A
- VERNA210059010010A
- VERNA200010005010A
- VERNA210025007510B
- Max. temperature:
- 260 °C
- 260 °C
- 327 °C
- 327 °C
- 260 °C
- 327 °C
- Min. temperature:
- −200 °C
- −200 °C
- −200 °C
- −200 °C
- −200 °C
- −200 °C