Anti-PDIA4 Rabbit Recombinant Antibody [clone: R02-1J3]
Catalog # ABNORAB02293.100UL
Supplier: Abnova
Specifications
- Antigen symbol:PDIA4
- Clonality:Recombinant
- Clone:R02-1J3
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human PDIA4.
- Pk:100 µl