ContactUsLinkComponent
Tube holder S
Tube holder S
  BINH1014525
undefined
Tube holder S
  BINH1014525
  • Description:
    Tube Holder S
  • Pk:
    1
Tube holder to support all S series electron tubes for easy and safe operation. The five pin sockets for the tube are concealed inside the tube holder. A cathode protection switch is integrated into the tube holder, to protect the heated cathode from excessive voltage. The base plate has a slot for attaching the Helmholtz pair of coils S.
  • Terminals : 4 mm safety sockets
  • Dimensions: approx. 130×190×250 mm³
  • Weight: approx. 570 g