ContactUsLinkComponent
Vertical electrophoresis system, MV10
Vertical electrophoresis system, MV10
Catalog # 732-1427
Supplier:  EDVOTEK
undefined
Vertical electrophoresis system, MV10
Catalog # 732-1427
Supplier:  EDVOTEK
Specifications
  • Description:
    Vertical electrophoresis system, MV10
  • Pk:
    1
Specifications
About this item
Designed for separation of proteins on polyacrylamide gels, the MV10 unit holds one 9×19 cm gel cassette and can accommodate most pre-cast or self-made gels.
  • Holds one gel cassette
  • All platinum electrodes
  • Safety interlock cover
  • Safety electrical leads