Orders LinkContactUsLinkComponent
PCR beads
PCR Beads, 1
Catalog # EDVO625
Supplier:   EDVOTEK
PCR beads
Catalog # EDVO625
Supplier:  EDVOTEK
CAS Number:  
PCR beads
PCR Beads, 1
CAS Number:  
Price
60,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Each PCR Bead contains Taq DNA polymerase, Taq DNA polymerase buffer, dNTP mixture and MgCl₂. Pack contains sufficient for 25 reactions.
Specifications
Specifications
  • Pk:
    1
  • Description:
    PCR Beads