L-MARK® LC Autosampler Syringes
Supplier: Trajan Scientific and Medical
Certificates
About this item
The comprehensive range of L-MARK® Gold and L-MARK syringes for LC systems are designed and tested to meet critical instrument parameters.
- Plunger guide maintains optimal plunger alignment for extended plunger lifetime
- Syringes available with polyethylene, PTFE and black PTFE tipped plungers
- Titanium plungers for longer lifetime
- Needle setting in the syringe barrel is protected by a PTFE barrier for reduced exposure of the adhesive to samples
- The L-MARK Gold syringes include a chemically inert needle for extremely low carryover
Accuracy and reproducibility: ±1% (dispensed volume)
Temperature range: 5 to 70 °C
Specifications
- Catalog No:
- SGEALMK.2620517BL
- SGEALMK.3030095
- SGEALMK2620616C-6P
- SGEALMK2620821C-6P
- SGEALMK2620721C-6P
- SGEALMK2606416C-6P
- SGEALMK2606416B-6P
- SGEAKLMK.26208212P
- SGEALMK.2616822
- SGEALMK.2620820
- SGEALMK.2620821-6P
- SGEALMK.2620720-2P
- SGEALMK.2620517C
- SGEALMK.2616422
- SGEALMK.2620718
- SGEALMK.2620518-6P
- SGEALMK.2620614C
- SGEALMK.3030029
- SGEALMK.2620719C
- SGEALMK.3030021
- SGEALMK2620517C-6P
- SGEALMK.2620617-2P
- SGEALMK.2620615-6P
- SGEALMK2620517C-BL
- SGEALMK.3030022
- SGEALMK.2620835-BL
- SGEALMK.2620517-2P
- SGEALMK.3030031
- SGEALMK.2620615
- SGEALMK.2620820-2P
- SGEALMK.2620720C
- SGEALMK.2616704C
- SGEALMK.2606416-6P
- SGEALMK.2620718C
- SGEALMK.2620518C
- SGEALMK.2620718-2P
- SGEALMK.2620080-BL
- SGEALMK.2620518
- SGEALMK.2620521C
- SGEALMK.2620718-6P
- SGEALMK.2620616-6P
- SGEALMK2620615C-BL
- SGEALMK.2620721-6P
- SGEALMK.2620719-2P
- SGEALMK.2620935-BL
- SGEALMK.2620719-6P
- SGEALMK2620719C-2P
- SGEALMK.2620616
- SGEALMK2620719C-BL
- SGEALMK.3030908
- SGEALMK.2620614
- SGEALMK.2620615-2P
- SGEALMK.2620617-6P
- SGEALMK.2620517-6P
- SGEALMK.2606416
- SGEALMK.2620517
- SGEALMK.2620719
- SGEALMK.2616502C
- SGEALMK.2620720-6P
- SGEALMK.2620719-BL
- SGEALMK.2620720
- SGEALMK.2620616-2P
- SGEALMK.2620721-2P
- SGEALMK2620719C-6P
- SGEALMK2620617C-BL
- SGEALMK2620719B-6P
- SGEALMK2620721C-BL
- SGEAKLMK.2620821
- SGEALMK.2620721C
- SGEALMK.2620615C
- SGEALMK.2616922
- SGEALMK.2620721
- SGEALMK.2620820-6P
- SGEALMK.2620740
- SGEALMK.2620614-2P
- SGEALMK.2606416-2P
- SGEALMK.2620617
- SGEALMK.2620735-BL
- SGEALMK.2620617C
- SGEALMK.2606416C
- Needle length:
- 51 mm
- 57 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 57 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 57 mm
- 51 mm
- 51 mm
- 51 mm
- 57 mm
- 51 mm
- 57 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 85 mm
- 51 mm
- 57 mm
- 51 mm
- 51 mm
- 51 mm
- 57 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 85 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 57 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 57 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm