ContactUsLinkComponent
Scotch-Brite™ 3000 Diamond Floor Brush
Scotch-Brite™ 3000 Diamond Floor Brush
Catalog # MMMA7100262371
Supplier:  3M
Scotch-Brite™ 3000 Diamond Floor Brush
Catalog # MMMA7100262371
Supplier:  3M
Specifications
  • Ø:
    431,8 mm
  • For:
    Floor, polished concrete
  • Description:
    Floor brush, round, 3000 grit, 175 - 600 min⁻¹ machine speed
  • Pk:
    2
Specifications
About this item
Scotch-Brite™ Diamond floor brush 3000 series is designed to clean and polish uncoated concrete, terrazzo and potentially other stone floors in one step. Brush 3000 is a finer grit brush intended for daily use on floors where medium to high gloss is desired.
  • Provides both cleaning and gloss recovery in one step
  • Conformable, flexible diamond bristles reach low spots in floors
  • Longer life and durability when compared to traditional and diamond floor pads
  • A more sustainable solution as they can be used with just water to clean and maintain floors
  • Longer brush life means less waste in landfills
  • Coating-free maintenance system - no need for chemicals for floor stripping or finishing
  • Brush can be used with just water, providing a more sustainable solution
  • Designed for daily use with water on a low-speed machine (175 to 600 min⁻¹)
It is conformable allowing the flexible bristles to reach low spots on the floor while minimising debris clogging. No assembly or pre-soaking is required and it is easy to install on standard, low-speed equipment.