96 Well Plate, RCH TRACE-N20
TCAT611-0101RCH
: Tecan
- Description:Plate RCH TRACE-N20, 1ml, 15 µm
- Environmentally Preferable:
- Phase:TRACE-N20
- Filling:10 mg
- Pk:1
An end-capped C18 silica sorbent ideal for reverse-phase retention of neutral and polar analytes.
- Reversed-phase
- Liquid handler compatible
A 15 µm particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Formulated for 25-OH-VitD/steroid assays.