Dosipet Dispensers
: KARTELL
Suitable for measuring out organic polar and non- polar solvents, including halogenated solvents and concentrated mineral acids, (including particularly volatile acids such as nitric and trifluoroacetic acids).
- Can be used with peroxides, hydrochloric acid, and sulphuric acid
- Borosilicate glass piston
- Reagents come into contact only with parts made of PTFE
Available in multiple versions, depending on the reagents to be dispensed. The Dosipet S is useful for dosing concentrated acids, polar and non-polar solvents. The Dosipet HF is specially developed for hydrofluoric acid dosing. They are supplied complete with tubes for filling/discharging, as well as with a set of adaptors.