CRS SPure™ H₂O Gas Filters
Supplier: Trajan Scientific and Medical
Certificates
About this item
Pure gas is a crictical requirement in gas chromatography, spectroscopy, optics, lithography and numerous other applications in manufacturing and laboratories. The SPure™ H₂O filters removes moisture, oil and dust from air and inert gases.
- High-strength polycarbonate and acrylic bodies with aluminum end caps
- Available with standard brass and stainless steel compression fittings
- It is suitable for general purpose, non-critical laboratory use
- Maximum pressure 6,89 bar
It is not recommended for house compressed air lines or for GC carrier gases and not use for with hazardous, flammable, or reactive gases.