SPE Column, NBE OFXQ
: Tecan
Proprietary blend of ion-exchange and water-wettable polymer targeted towards oral fluid collection devices.
- Ion-exchange
- Liquid handler compatible
A proprietary polymer resin blend of ion exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.