ZEN 3.9 Software for Microscopy Workstation
ZEN 3.9 software, Multi channels
Catalog # ZEIS4101350002390
Supplier: ZEISS
Jump to:
Product Details & Documents
ZEN 3.9 software is an image acquisition and processor under Windows 10×64 user interface configurable.
- Sheer control of the microscope systems and components
- Extensive acquisition and analysis and results are provided as CZI image format
It includes with following functions such as arivis cloud advanced, basic 3D viewing, colocalization, connect basic, direct processing, manual EDF, measurement, panorama, spectral unmixing, ZEN data storage client and acquisition base (time series and multichannel).
Specifications
- Description:ZEN 3.9 software
- Channels:Multi channels