Cerex® TRACE-J SPE Columns
Supplier: Tecan
Certificates
About this item
Strong cationic columns with small particle and pore sizes for aggressive cleanup of basic compounds.
- Ion-exchange
- Liquid handler compatible
A 18 µm polymer micro particulate with strong cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Ideal for basic target compounds. Formulated for pain panel/FNT.