Order Entry
Germany
ContactUsLinkComponent
Visible Storage Cabinet, 12 Drawers
Visible Storage Cabinet, 12 Drawers
Catalog # MULC14211.294.3
Supplier:  MULTIROIR
undefined
Visible Storage Cabinet, 12 Drawers
Catalog # MULC14211.294.3
Supplier:  MULTIROIR

Specifications

  • Colour:
    Dark blue
  • Description:
    Visible storage cabinet, 12 drawers, interior dimension: 92W×59H mm
  • Material:
    PP
  • W×D×H:
    310×180×290 mm
  • Pk:
    1

Specifications

About this item