Centrifuge Tube Slicer
Catalog # BECM347960
Supplier: Beckman Coulter
Specifications
- Description:CentriTube slicer
- Pk:1
Specifications
About this item
This CentriTube slicer is used for slicing polypropylene, polycarbonate, and cellulose propionate centrifuge tubes with diameters of 5 to 13 mm.