jo_Clean Strap Mid, S3 ESD Safety Shoes, White
: ELTEN
Thanks to its excellent discharge capability, the shoe is suitable for
work in ESD sensitive or electrostatically protected areas (EPA).
- Full, padded bellows tongue
- MONO-PU sole jo_MONO
- Composite toe cap
- Metal- and leather-free equipment
- Available in white finish
Hydrophobised microfibre
Fabric lining for natural ventilation. Full-length inlay sole. Non-metallic, penetration-resistant midsole.
: EN ISO 20345 S3 SRC, form B, ESD Standard, The shoes comply to the standard 61340-5-1, Certification in accordance with DGUV rule 112-191.
- Catalog No:
- ELTN16681/39
- ELTN16681/41
- ELTN16681/48
- ELTN16681/40
- ELTN16681/43
- ELTN16681/42
- ELTN16681/45
- ELTN16681/44
- ELTN16681/36
- ELTN16681/47
- ELTN16681/46
- ELTN16681/38
- ELTN16681/37
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001