ContactUsLinkComponent
Sentinel® Quick-Loc Belt
Sentinel® Quick-Loc Belt
Catalog # ILCDS-4011
Supplier:  ILC DOVER
undefined
Sentinel® Quick-Loc Belt
Catalog # ILCDS-4011
Supplier:  ILC DOVER
Specifications
  • Description:
    Quick-Loc waist belt
  • Seq:
    05
  • Pk:
    1
Specifications
About this item
Easy to clean waist belt quickly and securely snaps into the ergonomically designed Sentinel® blowers for hands-free operation.
  • Blower quickly snaps into the belt
  • Easily cleaned with wipes or immersion
Belt is designed to contour to the user's back for increased comfort.