Micrometer Film Applicators
Adjustable Micrometer Film Applicator. The applicator blade can be adjusted in 10 μm increments by means of 2 μm heads, over a gap size from 0 to 8 mm.
- Stainless steel knife blade with precision ground edge
- Wet film thickness adjustable to 8 mm maximum
- Micrometer head accuracy is ±10 µm
- Ergonomic design facilitates fast accurate draw downs
- Anodised aluminium material, through hardened medical grade stainless steel
- Black anodised aluminium frame provides a large paint well
The applicator offers the user flexibility to adjust the optimum gap size for the sample application. Ideally suited to research projects where small variations in film thickness need to be evaluated accurately.
: ASTM D 823.
: Micrometer film applicator, 1,5 mm Allen key supplied.