Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boxes
Storage Boxes
Accessories for SAN 400E Dividable Storage Box
Accessories for SAN 400E Dividable Storage Box
Accessories for SAN 400E Dividable Storage Box
Catalog # MULC50209SEPELB91
Supplier:
MULTIROIR
Expand image
Accessories for SAN 400E Dividable Storage Box
Catalog # MULC50209SEPELB91
Supplier:
MULTIROIR
Specifications
Description:
Set of 10 Separations SEP 91
Pk:
1
Specifications
About this item
Product Details
Product Details