ContactUsLinkComponent
Remote Control, WiCo T 25 easy clean
Remote Control, WiCo T 25 easy clean
Catalog # IKAA25006310
Supplier:  IKA
Remote Control, WiCo T 25 easy clean
Catalog # IKAA25006310
Supplier:  IKA
Specifications
  • Description:
    WiCo T 25 easy clean remote control
  • Pk:
    1
  • For:
    T 25 easy clean control only (not compatible with other models)
Specifications
About this item
The WiCo wireless remote control allows user to operate and monitor the disperser T 25 easy clean control e.g. in a fume hood and therefore helps protect the user from exposure to toxic materials.
  • WiCo is only compatible with T 25 easy clean control disperser
Delivery: Supplied with wall mount WH 1 1 as well as two USB cables.