Order Entry
Germany
ContactUsLinkComponent
Elix® Electrodeionisation (EDI) Module 40 L/h
Catalog # MILLZLXM0D040
Supplier:  Merck
Elix® Electrodeionisation (EDI) Module 40 L/h
Catalog # MILLZLXM0D040
Supplier:  Merck

Specifications

  • Description:
    Elix® Electrodeionisation (EDI), module 40 L/h
  • Pk:
    1

Specifications

About this item

Deionises and produces superior quality Type 2 pure water, flow rate of 40 L/h. Eco-friendly EDI module eliminates the need for chemical regeneration.

Following reverse osmosis (RO) treatment, the Elix® Electrodeionisation (EDI) Module continuously deionises water to produce high-quality pure water, without any maintenance at a flow rate of 40 L/h. This constant-quality Type 2 pure water is suitable for critical applications, instrument feed and general laboratory use. The eco-friendly EDI module eliminates the need for hazardous chemical regeneration procedures, replacement of costly resins, and changing DI cartridges.

Components: Mixed bed ion-exchange resin

Application: Deionises and produces superior quality Type 2 pure water • Flow rate of 40 L/h • Eco-friendly EDI module eliminates the need for chemical regeneration.