Human Recombinant CDC2 (His Tag) (from E. coli)
Catalog # NOVUNBP2-52017-0.1
Supplier: Novus Biologicals
CAS Number:
Specifications
- Pk:0,1 mg
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Gene ID:CDK1
- Amino acid number:MGSSHHHHHH SSGLVPRGSH MEDYTKIEKI GEGTYGVVYK GRHKTTGQVV AMKKIRLESE EEGVPSTAIR EISLLKELRH PNIVSLQDVL MQDSRLYLIF EFLSMDLKKY LDSIPPGQYM DSSLVKSYLY QILQGIVFCH SRRVLHRDLK PQNLLIDDKG TIKLADFGLA RAFGIPIRVY THEVVTLWYR SPEVLLGSAR YSTPVDIWSI GTIFAELATK KPLFHGDSEI DQLFRIFRAL GTPNNEVWPE VESLQDYKNT FPKWKPGSLA SHVKNLDENG LDLLSKMLIY DPAKRISGKM ALNHPYFNDL DNQIKKM
- Protein synonyms:EC 2.7.11.23|CDK1|p34 protein kinase|Cell division control protein 2 homolog|P34CDC2|cyclin-dependent kinase 1|CDC28A|EC 2.7.11.22|G1 to S and G2 to M|Cell division protein kinase 1|CDC2MGC111195|cell cycle controller CDC2|DKFZp686L20222
- Protein/peptide name:CDC2
- Purity:>85% as determined by SDS-PAGE.
- Molecular weight:34,9 kDa
- Concentration:1 mg/ml
Specifications
About this item
A denatured recombinant protein with a N-terminal his-tag and corresponding to the amino acids 1 - 297 of human CDC2/CDK1.