Orders LinkContactUsLinkComponent
Microscope, Primostar 3
Microscope, Primostar 3
Primostar 3 microscope
Catalog # ZEIS4155010031000
Supplier:   ZEISS
Microscope, Primostar 3
Catalog # ZEIS4155010031000
Supplier:  ZEISS
Microscope, Primostar 3
Primostar 3 microscope
Price
3 177,39 €
Log in for availability
1 * 1 ST
Product Details & Documents
Primostar 3 microscope is a robust and compact microscope for digital teaching and routine lab work.
Transmitted light brightfield, darkfield, simple polarisation, phase contrast and fluorescence contrast are contrast techniques of choice.
Use free of charge labscope app on your Windows PC or iOS device to create images, movies.
Specifications
  • Description:
    Primostar 3 microscope
  • Cat. No.:
    ZEIS4155010031000
  • Condenser:
    ABBE condenser 0,9
  • Nosepiece:
    Quintuple, reversed
  • Model:
    Primostar 3
  • Objectives:
    iPlan-Achromat 4×, 10×, 40× Ph2
  • Display:
    LED
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Micrometer for Primostar 3, 10:100, 25 mm
    Micrometer for Primostar 3, 10:100, 25 mm
    ZEIS4155011815000
    Log in for availability
    1 * 1 ST - 61,88 €
Product Information
  • Catalog #
    Working distance
    Availability
    Price
  • iPlan achromat objective lens, 10×/0,25 Ph1
    iPlan achromat objective lens, 10×/0,25 Ph1
    ZEIS4155011615000
    18,3 m
    Log in for availability
    1 * 1 ST - 308,00 €
Product Information
  • Catalog #
    Working distance
    Availability
    Price
  • iPlan achromat objective lens, 100×/1,25 oil
    iPlan achromat objective lens, 100×/1,25 oil
    ZEIS4155011641000
    0,16 m
    Log in for availability
    1 * 1 ST - 394,64 €
Product Information
  • Catalog #
    Working distance
    Availability
    Price
  • iPlan achromat objective lens, 20×/0,45 Ph2
    iPlan achromat objective lens, 20×/0,45 Ph2
    ZEIS4155011625000
    0,9 m
    Log in for availability
    1 * 1 ST - 367,25 €
Product Information
  • Catalog #
    Working distance
    Availability
    Price
  • iPlan achromat objective lens, 20×/0,45
    iPlan achromat objective lens, 20×/0,45
    ZEIS4155011622000
    0,9 m
    Log in for availability
    1 * 1 ST - 242,46 €
Product Information
  • Catalog #
    Working distance
    Availability
    Price
  • iPlan achromat objective lens, 40×/0,65 Ph2
    iPlan achromat objective lens, 40×/0,65 Ph2
    ZEIS4155011635000
    0,6 m
    Log in for availability
    1 * 1 ST - 394,64 €
Product Information
  • Catalog #
    Working distance
    Availability
    Price
  • iPlan achromat objective lens, 4×/0,10
    iPlan achromat objective lens, 4×/0,10
    ZEIS4155011600000
    30,7 m
    Log in for availability
    1 * 1 ST - 136,00 €
Product Information
  • Catalog #
    Working distance
    Availability
    Price
  • iPlan achromat objective lens, 10×/0,25
    iPlan achromat objective lens, 10×/0,25
    ZEIS4155011610000
    17,4 m
    Log in for availability
    1 * 1 ST - 167,39 €