ContactUsLinkComponent
Real Time RT-PCR Kit
Real Time RT-PCR Kit
Catalog # GSDCPCOV6111
CAS Number:  
Real Time RT-PCR Kit
Catalog # GSDCPCOV6111
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    GSD NovaPrime® TSP SARS-CoV-2, 1×96 reactions
Specifications
About this item
High quality RT-PCR kits for detection of various infectious diseases.
  • Fast and reliable results in approximately 1 hour
  • Validated for a variety of specimen types
  • Excellent performance with high sensitivity and specificity
  • Extraction/inhibition and positive control included
  • Suitable for most PCR platforms