Specifications
- Colour:Grey
- Description:Vinyl anti-skid cleanroom tape
- Length:18.28 m
- Pk:1 Roll
Specifications
About this item
UltraTape vinyl anti-skid cleanroom tape is a high-quality tape designed for use in cleanroom environments where safety and contamination control are critical. This tape is made from a durable vinyl material that is specially formulated to provide excellent adhesion and resistance to skidding, making it ideal for use in areas where slipping hazards are a concern.
- With its low particulate and ionic contamination
- Excellent choice for use in semiconductor and electronics manufacturing
- Used in pharmaceutical and biotech applications
Its high-visibility yellow color makes it easy to identify and locate, even in low-light conditions. The tape is easy to apply and remove, and its strong adhesive ensures that it stays in place even under heavy foot traffic.
Cleanroom Tape is also resistant to most chemicals and solvents, making it an ideal choice for use in harsh industrial environments. Its durable construction allows it to withstand frequent cleaning and sterilisation, making it a cost-effective solution for maintaining a safe and clean work environment.
Tape is a reliable and high-quality tape that provides excellent adhesion, resistance to skidding, and durability, making it an ideal choice for use in a wide range of cleanroom and industrial applications where safety and contamination control are critical.