Slide Closure Zip Lock Bags, Sterile
Catalog # KACC1215SZLDPST2
Supplier: KEYSTONE CLEANROOM PRODUCTS
Specifications
- Colour:Clear
- Description:Slide closure zip lock bag, sterile, 2 per pack, double pouched with hole in pouch corner
- L×W:380×305 mm
- Pk:200
Specifications
About this item
Pre-sterilised and packaged in multiple layers of Tyvek peel pouches and poly bags to gain access into the aseptic area without excessive spraying and wiping with sanitisation agents. The bags are used for the removal and transport of media plates from the aseptic area to QC laboratories.
- Manufactured and packaged in an ISO certified cleanroom
- Tear and puncture resistant 2,7 mill low density polyethylene
- Each bag has an integral zip seal with the slide lock closure system
- Two zip-lock bags are double packaged in Tyvek/film chevron peel pouches and then inside a third, sealed poly bag layer
- Labelled with lot number, date of manufacture and expiration date
- Validated sterile to a 10⁻⁶ SAL
- Certificates of conformance and sterility sent with each shipment
Environmental monitoring technicians can confidently remove media plates from the aseptic area and transport them to QC laboratories while reducing the risks of false positives by using Keystone Cleanroom Products pre-sterilised slide-lock closure zip-lock bags.
Certifications: CoC and certificate of sterility.