Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
AmberChrom™ hydrogen form
AmberChrom™ hydrogen form 50W X8 16-50 mesh, strongly acidic, Supelco®
AmberChrom™ hydrogen form 50W X8 16-50 mesh, strongly acidic, Supelco®
AmberChrom™ hydrogen form 50W X8 16-50 mesh, strongly acidic, Supelco®
Catalog # SIAL44504-100G
Supplier:
Merck
CAS Number:
Expand image
AmberChrom™ hydrogen form 50W X8 16-50 mesh, strongly acidic, Supelco®
Catalog # SIAL44504-100G
Supplier:
Merck
CAS Number:
Specifications
Pk:
100 g
Seq:
3401
Specifications
About this item
Product Details
:
CAS Number:
69011-22-9
MDL Number:
MFCD00132722
Product Details