Order Entry
Germany
ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP) VLSI, Electronic Grade
Health
Irritant
N-Methyl-2-pyrrolidone (NMP) VLSI, Electronic Grade
Catalog # 50489118.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
N-Methyl-2-pyrrolidone (NMP) VLSI, Electronic Grade
Catalog # 50489118.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC):
    ≥ 99.5 %
  • Identification (IR):
    Passes test
  • Colouration:
    ≤ 50 APHA
  • Particle size(>0.50 μm)(/ml)(at filling):
    ≤ 80
  • Resistivity (MOhm-cm):
    ≥ 1
  • Free acid (as C₂H₅COOH):
    ≤ 50 ppm
  • Free amine (as methylamine):
    ≤ 20 ppm
  • Water:
    ≤ 1000 ppm
  • Cl (Chloride):
    ≤ 500 ppb
  • PO₄ (Phosphate):
    ≤ 1000 ppb
  • SO₄ (Sulphate):
    ≤ 500 ppb
  • Ag (Silver):
    ≤ 10 ppb
  • Al (Aluminium):
    ≤ 50 ppb
  • As (Arsenic):
    ≤ 10 ppb
  • Au (Gold):
    ≤ 10 ppb
  • B (Boron):
    ≤ 10 ppb
  • Ba (Barium):
    ≤ 50 ppb
  • Be (Beryllium):
    ≤ 10 ppb
  • Bi (Bismuth):
    ≤ 10 ppb
  • Ca (Calcium):
    ≤ 50 ppb
  • Cd (Cadmium):
    ≤ 10 ppb
  • Co (Cobalt):
    ≤ 10 ppb
  • Cr (Chromium):
    ≤ 10 ppb
  • Cu (Copper):
    ≤ 10 ppb
  • Fe (Iron):
    ≤ 20 ppb
  • Ga (Gallium):
    ≤ 10 ppb
  • Ge (Germanium):
    ≤ 10 ppb
  • In (Indium):
    ≤ 10 ppb
  • K (Potassium):
    ≤ 50 ppb
  • Li (Lithium):
    ≤ 10 ppb
  • Mg (Magnesium):
    ≤ 20 ppb
  • Mn (Manganese):
    ≤ 10 ppb
  • Mo (Molybdenum):
    ≤ 20 ppb
  • Na (Sodium):
    ≤ 100 ppb
  • Ni (Nickel):
    ≤ 5 ppb
  • Pb (Lead):
    ≤ 10 ppb
  • Pt (Platinum):
    ≤ 5 ppb
  • Sb (Antimony):
    ≤ 5 ppb
  • Sn (Tin):
    ≤ 10 ppb
  • Sr (Strontium):
    ≤ 5 ppb
  • Ti (Titanium):
    ≤ 10 ppb
  • Tl (Thallium):
    ≤ 10 ppb
  • V (Vanadium):
    ≤ 10 ppb
  • Zn (Zinc):
    ≤ 10 ppb
  • Zr (Zirconium):
    ≤ 10 ppb

Specification Test Results

About this item

 :
 :99.13 g/mol
Merck Index:13,06140
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
Flash Pt:91 °C
Storage Temperature:Ambient
EINECS:212-828-1
REACH:01-2119472430-46
 :GHS08
 :GHS07
CAS Number:872-50-4
Formula:C₅H₉NO
MDL Number:MFCD00003193