ContactUsLinkComponent
TELSTAR Biological Safety Cabinets, Class I, BiOptima ONE
TELSTAR Biological Safety Cabinets, Class I, BiOptima ONE
Certificates
undefined
TELSTAR Biological Safety Cabinets, Class I, BiOptima ONE
Specifications
  • Catalog No:
  • TELR1775507
  • TELR1776556
  • Filters:
  • HEPA H14 (G3 prefilter)
  • HEPA H14 (G3 prefilter)
  • Noise level:
  • <58
  • <55
  • Electrical:
  • 230 V, 50 Hz, 190 W
  • 230 V, 50 Hz, 150 W
  • Ext. W×D×H:
  • 1338×797×1450 mm
  • 1033×797×1450 mm
  • Int. W×D×H:
  • 1190×548×666 mm
  • 885×548×666 mm
  • Weight:
  • 210 kg
  • 180 kg
Specifications