Order Entry
Germany
ContactUsLinkComponent
 
XELA UHPLC Columns, C8
 :  ISERA
 
XELA UHPLC Columns, C8
 :  ISERA

 

  • Catalog No:
  • ISERA211-D11V00
  • ISERA212-D11A30
  • ISERA211-D11M00
  • ISERA211-D11U00
  • ISERA211-D11H00
  • ISERA211-D11K00
  • ISERA211-D11F00
  • ISERA211-D11G00
  • ISERA211-D11B00
  • ISERA211-D11D00
  • ISERA211-D11X00
  • ISERA211-D11A00
  • ISERA211-D11Z00
  • Description:
  • Analytical columns
  • Guard column
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Analytical columns
  • Phase name:
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • Endcapped:
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • High
  • Max. pressure:
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • pH range:
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • USP code:
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • Storage:
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • Shipping conditions:
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.

 

Guard column holders