Order Entry
Germany
ContactUsLinkComponent
XELA UHPLC Columns, Silica
XELA UHPLC Columns, Silica
Supplier:  ISERA
Certificates
XELA UHPLC Columns, Silica
Supplier:  ISERA

Specifications

  • Catalog No:
  • ISERA212-411A30
  • ISERA211-411D00
  • ISERA211-411A00
  • ISERA211-411Z00
  • ISERA211-411B00
  • ISERA211-411V00
  • ISERA211-411X00
  • ISERA211-411M00
  • ISERA211-411U00
  • ISERA211-411H00
  • ISERA211-411K00
  • ISERA211-411F00
  • ISERA211-411G00
  • Description:
  • Guard column
  • Analytical column
  • Analytical column
  • Analytical column
  • Analytical column
  • Analytical column
  • Analytical column
  • Analytical column
  • Analytical column
  • Analytical column
  • Analytical column
  • Analytical column
  • Analytical column
  • Phase name:
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • Max. pressure:
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • pH range:
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • USP code:
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • Storage:
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • Shipping conditions:
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.

Specifications

Guard column holders

Product Information