Oil Sorbent Pillows
Oil sorbent pillows, T-30, 180×380 mm
Catalog # MMMAT3016
Supplier: 3M
Jump to:
Product Details & Documents
3M™ Oil sorbent pillows are open mesh bags designed with a scrim that encases particulate sorbent. Excellent for handling large quantities of light oil in confined spaces, they are UV-resistant and will not deteriorate or sink.
- Ideal for absorbing oil from areas where water may also be present
- Open mesh bags are tear-resistant and feature a high capacity for holding liquid
- Ideal for placing at the source of a leak
Being hydrophobic repelling water, they are ideal for absorbing oil from areas where water may also be present.
Specifications
- Description:Oil sorbent pillows, T-30
- L×W:180×380 mm