ContactUsLinkComponent
Scotch-Brite™ 770 Cellulosic Sponge Laminate
Scotch-Brite™ 770 Cellulosic Sponge Laminate
Catalog # MMMASCH770
Supplier:  3M
undefined
Scotch-Brite™ 770 Cellulosic Sponge Laminate
Catalog # MMMASCH770
Supplier:  3M
Specifications
  • Colour:
    Blue
  • Description:
    Scotch-Brite™ Cellulosic sponge laminate, 770
  • L×W:
    131×88 mm
  • Pk:
    10
Specifications
About this item
Scotch-Brite™ Cellulosic sponge laminate 770 is a dual-function cleaning tool made from a combination of a non-woven tablecloth containing very light abrasive particles to a pure cellulose sponge.
  • Great cleaning performance
  • High absorption power
  • Does not scratch even the most sensitive surfaces
  • Works with hot water or diluted bleach
  • Excellent hydrophilic properties
Remove grease and food without leaving scratches with Scotch-Brite™ Cellulosic Sponge Laminate 770. Made from a combination of a non-woven tablecloth containing very light abrasive particles to a pure cellulose sponge. The fine texture of the synthetic fibres on the cloth makes it a medium buffer abrasive that that works with hot water or diluted bleach to remove grease and food without scratching.