Order Entry
Germany
Orders LinkContactUsLinkComponent
 

J.T.Baker® Cell Lysis Solution, Biotech Reagent

   Sustainable Options Available
Supplier:   Avantor
J.T.Baker® Cell Lysis Solution, Biotech Reagent
Supplier:  Avantor
   Sustainable Options Available
Certificates
J.T.Baker® Cell Lysis Solution, Biotech Reagent

5 Options Available Below

Product Details & Documents

J.T.Baker® Cell Lysis Solution Biotech Reagent is a cGMP pre-formulated and ready to use detergent optimized for the extraction and recovery of AAV particles from producer cell lines (HEK293 parental and suspension lines, SF9). J.T.Baker® Cell Lysis Solution is a proprietary blend of nonionic surfactants, free of octyl phenol ethoxylate, which demonstrates comparable performance industry standard detergents while addressing environmental concerns associated with REACH and downstream clearance concerns. Designed for Gene Therapy workflows, this formulation is easy to use and handle as it is ready-to-use, has low viscosity, does not require cold temperature shipping or storage, and is available in sizes to support processes from development to large scale commercial batches. Detail description provided in technical literature.

  • Comparable cell lysis performance to octyl phenol ethoxylate
  • Improved protection of viral particles
  • Easy to use and handle due to reduced foaming and viscosity and Room Termperature shipping and storage
  • 0.1 µm filtered solution

Proprietary formulation. Not hazardous according to regulation 29 CFR 1910.1200 (OSHA HCS)

Chemical Specifications of GMP Cell lysis solution:
GMP Manufactured Product
Refractive Index: 1.1-1.5
Appearance: Passes Test
pH : For information only
Viscosity (25°C) : For information only
Density: 0.9 – 1.2 g/mL
Total Aerobic Microbial Count: 100 cfu/mL
Total Yeast and Mold Count: 10 cfu/mL
UV-Vis (max Absorption) at 380-450 nm: For information only

Chemical Specifications of GMP Cell lysis solution
GMP Manufactured Product
Refractive Index: 1.1-1.5
Appearance: Passes Test
pH: For information only
Viscosity (25°C): For information only
Density: 0.9 – 1.2 g/mL
Total Aerobic Microbial Count: 100 cfu/mL
Total Yeast and Mold Count: 10 cfu/mL
UV-Vis (max Absorption) at 380-450 nm: For information only

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • G193-01
  • G193-04
  • G193-05
  • G193-07
  • G193-09
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Density:
  • 0.9-1.2 g/ml
  • 0.9-1.2 g/ml
  • 0.9-1.2 g/ml
  • 0.9-1.2 g/ml
  • 0.9-1.2 g/ml
  • Environmental certification:
  • EPA safer choice chemical
  • EPA safer choice chemical
  • EPA safer choice chemical
  • EPA safer choice chemical
  • EPA safer choice chemical

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • G193-01
    Cell Lysis Solution, Biotech Reagent GMP
    Cell Lysis Solution, Biotech Reagent GMP
    Plastic bottle
    10 mL
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - 227,63 €
    227,63 €
    1 * 10 mL
       227,63 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    G193-01
    Environmentally Preferable
    Safer Human and Environmental Health
    Density
    0.9-1.2 g/ml
    Environmental certification
    EPA safer choice chemical
      Safer Human and Environmental Health
  • G193-04
    Cell Lysis Solution, Biotech Reagent GMP
    Cell Lysis Solution, Biotech Reagent GMP
    Plastic bottle
    200 mL
    Log in for availability
    Log in for availability
    1 * 200 mL
    1 * 200 mL - 1 879,00 €
    1 879,00 €
    1 * 200 mL
       1 879,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    G193-04
    Environmentally Preferable
    Safer Human and Environmental Health
    Density
    0.9-1.2 g/ml
    Environmental certification
    EPA safer choice chemical
      Safer Human and Environmental Health
  • G193-05
    Cell Lysis Solution, Biotech Reagent GMP
    Cell Lysis Solution, Biotech Reagent GMP
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 2 549,00 €
    2 549,00 €
    1 * 500 mL
       2 549,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    G193-05
    Environmentally Preferable
    Safer Human and Environmental Health
    Density
    0.9-1.2 g/ml
    Environmental certification
    EPA safer choice chemical
      Safer Human and Environmental Health
  • G193-07
    Cell Lysis Solution, Biotech Reagent GMP
    Cell Lysis Solution, Biotech Reagent GMP
    Plastic bottle
    2.000 mL
    Log in for availability
    Log in for availability
    1 * 2.000 mL
    1 * 2.000 mL - 9 529,00 €
    9 529,00 €
    1 * 2.000 mL
       9 529,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    G193-07
    Environmentally Preferable
    Safer Human and Environmental Health
    Density
    0.9-1.2 g/ml
    Environmental certification
    EPA safer choice chemical
      Safer Human and Environmental Health
  • G193-09
    Cell Lysis Solution, Biotech Reagent GMP
    Cell Lysis Solution, Biotech Reagent GMP
    Plastic bottle
    5.000 mL
    Log in for availability
    Log in for availability
    1 * 5.000 mL
    1 * 5.000 mL - 19 009,00 €
    19 009,00 €
    1 * 5.000 mL
       19 009,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    G193-09
    Environmentally Preferable
    Safer Human and Environmental Health
    Density
    0.9-1.2 g/ml
    Environmental certification
    EPA safer choice chemical
      Safer Human and Environmental Health